It acts on type I calcium/calmodulin dependent phosphodiesterase and also a useful electron donor. Phenothiazine finds uses in metal free organic dye sensitizers, dyes and antioxidants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It acts on type I calcium/calmodulin dependent phosphodiesterase and also a useful electron donor. Phenothiazine finds uses in metal free organic dye sensitizers, dyes and antioxidants.
Solubility
Soluble in water (0.13 mg/ml at 23°C), ethanol, acetone, and ether. Insoluble in chloroform.
Notes
Store under dry inert gas. Protect from sunlight. Ensure good ventilation.
RUO – Research Use Only
A S Horn.; S H Snyder. Chlorpromazine and dopamine: conformational similarities that correlate with the antischizophrenic activity of phenothiazine drugs. PNAS. 1971, 68 (10),2325-2328.
Lea Assed Bezerra Silva et. al. Antimicrobial Photodynamic Therapy for the Treatment of Teeth with Apical Periodontitis: A Histopathological Evaluation. Journal of Endodontics. 2012, 38 (3),360-366.
Lithiation in THF gives orange crystals of (phenothiazin-10-yl)lithium.3THF. Carboxylation and the orientation of a second lithiation have been studied: Angew. Chem. Int. Ed., 34, 921 (1995).