Thermo Scientific Chemicals

4-Bromobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: A12518.36
500 g, Each
Thermo Scientific Chemicals

4-Bromobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: A12518.36
500 g, Each
Quantity
Catalog number: A12518.36
also known as A12518-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
586-76-5
IUPAC Name
4-bromobenzoic acid
Molecular Formula
C7H5BrO2
InChI Key
TUXYZHVUPGXXQG-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(Br)C=C1
Form
Powder or crystalline powder
Melting Point (clear melt)
248-259°C
Appearance (Color)
White to pale cream or pale yellow
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Identification (FTIR)
Conforms

Description

4-Bromobenzoic acid was used to study the metabolic fate of 2-,3-and 4-bromo benzoic acids in rat hepatocytes incubation using high temperature liquid chromatography. It was used in bromine-specific detection of the metabolites of 2-,3-and 4-bromobenzoic acid in the urine and bile of rats by inductively coupled plasma mass spectrometry. It is also used as an intermediate for the synthesis of agrochemicals, pharmaceutical, chemical intermediates and liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzoic acid was used to study the metabolic fate of 2-,3-and 4-bromo benzoic acids in rat hepatocytes incubation using high temperature liquid chromatography. It was used in bromine-specific detection of the metabolites of 2-,3-and 4-bromobenzoic acid in the urine and bile of rats by inductively coupled plasma mass spectrometry. It is also used as an intermediate for the synthesis of agrochemicals, pharmaceutical, chemical intermediates and liquid crystals.

Solubility
Slightly soluble in hot water. Soluble in alcohol and ether.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text