beta-D-Glucose pentaacetate, 98%, Thermo Scientific Chemicals
beta-D-Glucose pentaacetate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

beta-D-Glucose pentaacetate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1000 g
Catalog number A12526.30
also known as A12526-30
Price (USD)/ Each
142.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
142.00
Add to cart
beta-D-Glucose pentaacetate, 98%, Thermo Scientific Chemicals
Catalog numberA12526.30
Price (USD)/ Each
142.00
-
Add to cart
Chemical Identifiers
CAS604-69-3
IUPAC Name[3,4,5,6-tetrakis(acetyloxy)oxan-2-yl]methyl acetate
Molecular FormulaC16H22O11
InChI KeyLPTITAGPBXDDGR-UHFFFAOYNA-N
SMILESCC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)White to pale cream
FormPowder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)128-134?C
View more
β-D-Glucose Pentaacetate is used in biochemical reaction and also used as an active pharmaceutical intermediate. Further, it is used to stimulate insulin release in rat pancreatic islets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
β-D-Glucose Pentaacetate is used in biochemical reaction and also used as an active pharmaceutical intermediate. Further, it is used to stimulate insulin release in rat pancreatic islets.

Solubility
Soluble in chloroform and methanol. Insoluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Ali, T. H.; Tajuddin, H. A. B.; Hussen, R. S. D.; Heidelberg, T. Increased Emulsion Stability for Reverse Y-Shaped Sugar-Based Surfactants. J. Surfactants Deterg. 2015, 18 (5), 881-886.
  2. Lülsdorf, N.; Pitzler, C.; Biggel, M.; Martinez, R.; Vojcic, L.; Schwaneberg, U. A flow cytometer-based whole cell screening toolbox for directed hydrolase evolution through fluorescent hydrogels. Chem. Commun. 2015, 51 (41), 8679-8682.