Thermo Scientific Chemicals

Terephthalic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A12527.36
500 g, Each
Thermo Scientific Chemicals

Terephthalic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A12527.36
500 g, Each
Quantity
Catalog number: A12527.36
also known as A12527-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-21-0
IUPAC Name
benzene-1,4-dicarboxylic acid
Molecular Formula
C8H6O4
InChI Key
KKEYFWRCBNTPAC-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(C=C1)C(O)=O
Assay (Silylated GC)
≥98.0%
Identification (FTIR)
Conforms
Appearance (Color)
White
Form
Powder
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%

Description

Terephthalic acid is used in the metal organic frame works. Terephthalic acid is used primarily to make polyethylene terephthalate (PET) resins and fibers, polyester fibers, films, polyethylene terephthalate solid-state resins and polyethylene terephthalate engineering resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Terephthalic acid is used in the metal organic frame works. Terephthalic acid is used primarily to make polyethylene terephthalate (PET) resins and fibers, polyester fibers, films, polyethylene terephthalate solid-state resins and polyethylene terephthalate engineering resins.

Solubility
slightly soluble in water (0,017 g/L at 25°C)

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It should be stored and used in molten form.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text