Ethyl crotonate, 98%, Thermo Scientific Chemicals
Ethyl crotonate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl crotonate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
100 g
500 g
Catalog number A12529.0E
also known as A12529-0E
Price (USD)/ Each
485.00
-
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)/ Each
485.00
Add to cart
Ethyl crotonate, 98%, Thermo Scientific Chemicals
Catalog numberA12529.0E
Price (USD)/ Each
485.00
-
Add to cart
Chemical Identifiers
CAS623-70-1
IUPAC Nameethyl (2E)-but-2-enoate
Molecular FormulaC6H10O2
InChI KeyZFDIRQKJPRINOQ-HWKANZROSA-N
SMILESCCOC(=O)\C=C\C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.4220-1.4260 @ 20?C
Ethyl crotonate is used as a solvent for cellulose esters as well as a plasticizer for acrylic resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl crotonate is used as a solvent for cellulose esters as well as a plasticizer for acrylic resins.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. J.L.Herrmann; G.R.Kieczykowski; R.H.Schlessinger. Deconjugative alkylation of the enolate anion derived from ethyl crotonate. Tetrahedron Letters. 1973, 14, (26)2433-2436
  2. Jerry R.Mohrig; Sabrina S.Fu; Randall W.King; Ronald Warnet; Gary Gustafson. Stereochemistry of nucleophilic conjugate addition. Addition of ethanol-d and 2-methyl-2-propanethiol-d to ethyl crotonate. J. Am. Chem. Soc. 1990, 112, (9)3665-3667