Thermo Scientific Chemicals

(1-Pentyl)triphenylphosphonium bromide, 98%, Thermo Scientific Chemicals

Catalog number: A12534.36
500 g, Each
Thermo Scientific Chemicals

(1-Pentyl)triphenylphosphonium bromide, 98%, Thermo Scientific Chemicals

Catalog number: A12534.36
500 g, Each
Quantity
Catalog number: A12534.36
also known as A12534-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
21406-61-1
IUPAC Name
pentyltriphenylphosphanium bromide
Molecular Formula
C23H26BrP
InChI Key
VAUKWMSXUKODHR-UHFFFAOYSA-M
SMILES
[Br-].CCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White
Form
Crystalline powder
Assay (Titration ex Bromide)
>97.5 to <102.5%
Water Content (Karl Fischer Titration)
<1.0%

Description

It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediates.

Solubility
Partly miscible in water. Soluble in methanol and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text