Thermo Scientific Chemicals

N,N,N',N'-Tetramethylethylenediamine, 99%, Thermo Scientific Chemicals

Catalog number: A12536.0F
2500 mL, Each
Thermo Scientific Chemicals

N,N,N',N'-Tetramethylethylenediamine, 99%, Thermo Scientific Chemicals

Catalog number: A12536.0F
2500 mL, Each
Quantity
Catalog number: A12536.0F
also known as A12536-0F
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-18-9
IUPAC Name
[2-(dimethylamino)ethyl]dimethylamine
Molecular Formula
C6H16N2
InChI Key
KWYHDKDOAIKMQN-UHFFFAOYSA-N
SMILES
CN(C)CCN(C)C
Appearance (Color)
Clear to slightly hazy colorless to yellow
Form
Liquid
Assay (GC)
≥98.5% [UK]
Comment
Material sourced in either US or UK
Identification (FTIR)
Conforms [UK]

Description

N,N,N',N'-Tetramethylethylenediamine is used with ammonium persulfate to catalyze the polymerization of acrylamide and bis-acrylamide to get polyacrylamide gels. It is also used for the separation of proteins or nucleic acids. Further, it is employed as a ligand for metal ions like zinc and copper. It is actively involved in the formation of anionic organometallic complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N,N′,N′-Tetramethylethylenediamine is used with ammonium persulfate to catalyze the polymerization of acrylamide and bis-acrylamide to get polyacrylamide gels. It is also used for the separation of proteins or nucleic acids. Further, it is employed as a ligand for metal ions like zinc and copper. It is actively involved in the formation of anionic organometallic complex.

Solubility
Miscible with water.

Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, carbon dioxide and copper.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text