Thermo Scientific Chemicals

Phenoxyacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: A12541.36
500 g, Each
Thermo Scientific Chemicals

Phenoxyacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: A12541.36
500 g, Each
Quantity
Catalog number: A12541.36
also known as A12541-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
122-59-8
IUPAC Name
2-phenoxyacetic acid
Molecular Formula
C8H8O3
InChI Key
LCPDWSOZIOUXRV-UHFFFAOYSA-N
SMILES
OC(=O)COC1=CC=CC=C1
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Melting Point (clear melt)
97.0-103.0°C
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

Phenoxyacetic acid is a flavoring ingredient. Its photodegradation using titanium dioxide as photocatalyst has been studied. Selective separation of penicillin V from phenoxyacetic acid using liquid membranes consisting of 1,2-dichloroethane and Amberlite LA-2 as carrier has been studied.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenoxyacetic acid is a flavoring ingredient. Its photodegradation using titanium dioxide as photocatalyst has been studied. Selective separation of penicillin V from phenoxyacetic acid using liquid membranes consisting of 1,2-dichloroethane and Amberlite LA-2 as carrier has been studied.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text