3'-Chloropropiophenone has been used in the preparation of 1-(3-chlorophenyl)-1-phenyl-1-propanol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3′-Chloropropiophenone has been used in the preparation of 1-(3-chlorophenyl)-1-phenyl-1-propanol.
Solubility
Soluble in methanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Celina Garcia; Patrick J.Walsh. Highly enantioselective catalytic phenylation of ketones with a constrained geometry titanium catalyst. Organic Letters. 2003, 5, (20)3641-3644
Giovanni Fronza; Claudio Fuganti; Piero Grasselli; Andrea Mele. The mode of bakers’ yeast transformation of 3-chloropropiophenone and related ketones. Synthesis of (2S)-[2-2H]propiophenone, (R)-fluoxetine, and (R)- and (S)-fenfluramine. J. Org. Chem. 1991, 56, (21)6019-6023