Thermo Scientific Chemicals

3'-Chloropropiophenone, 98%, Thermo Scientific Chemicals

Catalog number: A12542.36
500 g, Each
Thermo Scientific Chemicals

3'-Chloropropiophenone, 98%, Thermo Scientific Chemicals

Catalog number: A12542.36
500 g, Each
Quantity
Catalog number: A12542.36
also known as A12542-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
34841-35-5
IUPAC Name
1-(3-chlorophenyl)propan-1-one
Molecular Formula
C9H9ClO
InChI Key
PQWGFUFROKIJBO-UHFFFAOYSA-N
SMILES
CCC(=O)C1=CC=CC(Cl)=C1
Proton NMR
To conform to structure
Appearance (Color)
White to cream
Form
Crystalline powder, granules or chunks
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms

Description

3'-Chloropropiophenone has been used in the preparation of 1-(3-chlorophenyl)-1-phenyl-1-propanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′-Chloropropiophenone has been used in the preparation of 1-(3-chlorophenyl)-1-phenyl-1-propanol.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text