Informational:Taking your work to the clinic? Check out CTS Synth-a-Freeze Medium »
Gibco™

Synth-a-Freeze™ Cryopreservation Medium

Catalog number: A1254201
Have Questions?
Gibco™

Synth-a-Freeze™ Cryopreservation Medium

Catalog number: A1254201
Have Questions?
Catalog Number
A1254201
Unit Size
50 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A125420150 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Synth-a-Freeze Cryopreservation Medium is a defined, ready-to-use, liquid cryopreservation medium that can be used with a broad pectrum of primary cells, as well as cell lines, and demonstrates improved cell viability and recovery after thawing. Synth-a-Freeze medium contains 10% dimethylsulfoxide (DMSO) without antibiotics, antimycotics, hormones, growth factors, serum, or protein and is buffered with HEPES and sodium bicarbonate.

Synth-a-Freeze medium is intended for freezing and storing a variety of mammalian cell types including human keratinocytes, embryonic stem cells, neural stem cells, and mesenchymal stem cells. It should not be used for melanocytes.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type
Keratinocyte Cell, Embryonic Stem Cell, Neural Stem Cell, Mesenchymal Stem Cell
Classification
Animal Origin-free
Form
Liquid
Product Type
Cryopreservation Media
Serum Level
Serum-free
Sterility
Sterile
With Additives
HEPES, Phenol Red, DMSO (10%)
Endotoxin Level
< 1 EU/ml
Product Line
Gibco™, Synth-a-Freeze™
Purity or Quality Grade
Qualified for Further Manufacturing
Quantity
50 mL
Shipping Condition
Dry Ice

Contents & Storage

Store in freezer (-5 to -20°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text