Thermo Scientific Chemicals

4-Nitrobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A12543.09
10 g, Each
Thermo Scientific Chemicals

4-Nitrobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A12543.09
10 g, Each
Quantity
Catalog number: A12543.09
also known as A12543-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
122-04-3
IUPAC Name
4-nitrobenzoyl chloride
Molecular Formula
C7H4ClNO3
InChI Key
SKDHHIUENRGTHK-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(C=C1)C(Cl)=O
Appearance (Color)
Pale yellow to yellow
Form
Crystalline powder or crystals or flakes
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
70-75?C

Description

4-Nitrobenzoyl chloride is used in the preparation of polysubstituted furanonaphthoquinoines. It is also involved in Michael addition, Henry reaction, O-alkylation and cycloaddition reactions. Further, it is employed as an intermediate in active pharmaceutical ingredient and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzoyl chloride is used in the preparation of polysubstituted furanonaphthoquinoines. It is also involved in Michael addition, Henry reaction, O-alkylation and cycloaddition reactions. Further, it is employed as an intermediate in active pharmaceutical ingredient and dyes.

Solubility
Soluble in terahydrofuran, dichloromethane, chloroform and pyridine.

Notes
Moisture sensitive. Incompatible with water, alcohols, oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text