Thermo Scientific Chemicals

N,N-Dimethyl-n-octylamine, 95%, Thermo Scientific Chemicals

Catalog number: A12546.30
250 g, Each
Thermo Scientific Chemicals

N,N-Dimethyl-n-octylamine, 95%, Thermo Scientific Chemicals

Catalog number: A12546.30
250 g, Each
Quantity
Catalog number: A12546.30
also known as A12546-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7378-99-6
IUPAC Name
dimethyl(octyl)amine
Molecular Formula
C10H23N
InChI Key
UQKAOOAFEFCDGT-UHFFFAOYSA-N
SMILES
CCCCCCCCN(C)C
Appearance (Color)
Clear to slightly hazy colorless
Form
Liquid
Assay from Supplier's CofA
≥94.0% (sum of C8 isomers/chains)
Comment
Hazyness may settle to a white sediment on standing.
Refractive Index
1.4220-1.4270 @ 20?C

Description

N,N-Dimethyl-n-octylamine is used as ion-pairing agent in the quantitative estimation of thiazinamium methylsulphate in a pharmaceutical preparation by HPLC. It acts as a counterion in mobile phase for separation of tetracycline, tetracycline analogs and their potential impurities by reversed-phase ion-pair chromatography. Further, it is used in the study of chiral separations by complexation with proteins in capillary zone electrophoresis. In addition to this, it is used as a mobile phase in direct resolution and quantitation of (R)- and (S)-disopyramide

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyl-n-octylamine is used as ion-pairing agent in the quantitative estimation of thiazinamium methylsulphate in a pharmaceutical preparation by HPLC. It acts as a counterion in mobile phase for separation of tetracycline, tetracycline analogs and their potential impurities by reversed-phase ion-pair chromatography. Further, it is used in the study of chiral separations by complexation with proteins in capillary zone electrophoresis. In addition to this, it is used as a mobile phase in direct resolution and quantitation of (R)- and (S)-disopyramide

Solubility
Slightly miscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents, strong acids and copper.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text