Thermo Scientific Chemicals

2'-Hydroxyacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: A12550.22
100 g, Each
Thermo Scientific Chemicals

2'-Hydroxyacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: A12550.22
100 g, Each
Quantity
Catalog number: A12550.22
also known as A12550-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
118-93-4
IUPAC Name
1-(2-hydroxyphenyl)ethan-1-one
Molecular Formula
C8H8O2
InChI Key
JECYUBVRTQDVAT-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=CC=C1O
Form
Liquid
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Refractive Index
1.5570-1.5610 @ 20?C
Appearance (Color)
Clear colorless to pale yellow to yellow to orange

Description

2'-Hydroxyacetophenone is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Hydroxyacetophenone is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text