Thermo Scientific Chemicals

(+)-Quinidine, Thermo Scientific Chemicals

Catalog number: A12559.14
25 g, Each
Thermo Scientific Chemicals

(+)-Quinidine, Thermo Scientific Chemicals

Catalog number: A12559.14
25 g, Each
Quantity
Catalog number: A12559.14
also known as A12559-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56-54-2
IUPAC Name
(S)-[(2R,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol
Molecular Formula
C20H24N2O2
InChI Key
LOUPRKONTZGTKE-LHHVKLHASA-N
SMILES
COC1=CC=C2N=CC=C([C@H](O)[C@H]3C[C@@H]4CCN3C[C@@H]4C=C)C2=C1
Form
Powder or crystalline powder
Identification (FTIR)
Conforms
Optical Rotation
+250° to +270° (c=1 in ethanol)
Appearance (Color)
White to cream to yellow to brown
Assay (Non-aqueous acid-base Titration)
≥94.0 to ≤106.0% (quinidine + dihydroquinidine)

Description

(+)-Quinidine is used to treat or prevent many types of irregular heartbeats (heart arrhythmias such as atrial fibrillation). It may also be used to treat malaria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in methanol, chloroform, alcohol (1 g/36ml), benzene and DMSO(100mM). Slightly soluble in ether and petroleum ether. Insoluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text