Hydroxylamine-O-sulfonic acid is used in the synthesis of quinolones for the benzodiazepine site of GABAA receptors. It is also used as a polymerization catalyst and an amination reagent. It is involved in the preparation of diazene by reacting with an alkali. It is also used in the inhibition of polychlorinated dibenzo-p-dioxins (PCDD) as well as (polychlorinated biphenyl) PCB formation in co-combustion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Hydroxylamine-O-sulfonic acid is used in the synthesis of quinolones for the benzodiazepine site of GABAA receptors. It is also used as a polymerization catalyst and an amination reagent. It is involved in the preparation of diazene by reacting with an alkali. It is also used in the inhibition of polychlorinated dibenzo-p-dioxins (PCDD) as well as (polychlorinated biphenyl) PCB formation in co-combustion.
Solubility
Soluble in water and methanol. Slightly soluble in ethanol. Insoluble in ether and chloroform.
Notes
Moisture sensitive and hygroscopic. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases, acid chlorides, acid anhydrides and corrodes metal.
RUO – Research Use Only
General References:
- Aminating agent for nucleophiles. For a review of chemistry, see: Org. Prep. Proced. Int., 14, 265 (1982).
- Primary and secondary amines are converted to hydrazines: J. Org. Chem., 14, 813 (1949); Chem. Ber., 92, 2521 (1959); pyridine to 1-aminopyridinium salts: Org. Synth. Coll., 5, 43 (1977); benzotriazole to the benzyne precursor 1-aminobenzotriazole: J. Chem. Soc., Chem. Commun., 192, 193 (1965); for preparation of the bis(aminotriazolobenzene), a precursor of 1,4-dibenzyne, see: Tetrahedron Lett., 25, 2073 (1984).
- Aromatic rings can be aminated in the presence of AlCl3: J. Am. Chem. Soc., 83, 221 (1961). Reaction is more successful with certain heterocycles, N,N'-dimethyluracil giving the 5-amino-derivative almost quantitatively: Tetrahedron Lett., 2751 (1973).
- Reaction with ketones results in oxime sulfates which undergo the Beckmann rearrangement to amides on heating: J. Org. Chem. USSR, 17, 2284 (1981); the conversion of cyclic ketones to lactams can conveniently be carried out as a one-step operation: Synthesis, 537 (1979); J. Org. Chem., 54, 4419 (1989); Org. Synth. Coll., 7, 254 (1988).
- Aldehydes are converted directly to nitriles: Tetrahedron Lett., 3187 (1974): Helv. Chim. Acta, 59, 2786 (1976).
- A one-pot synthesis of aryl or heteroaryl amines from carboxylic acids (as their chlorides) has been reported, equivalent to the Schmidt or Hofmann reactions: Synthesis, 1143 (1990).
- Can also provide a source of the reducing agent diimide: Liebigs Ann. Chem., 645, 1 (1962); 721, 240 (1969). For examples of the use of diimide in the reduction of alkenes, see Hydrazine monohydrate, A14005.
- With aqueous NaOH, hydrodeamination of primary amines to alkanes is possible for simple aliphatic amines, amino acids or even peptides: J. Am. Chem. Soc., 100, 341 (1978).