Thermo Scientific Chemicals

Dimethyl succinate, 98%, Thermo Scientific Chemicals

Catalog number: A12565.22
100 g, Each
Thermo Scientific Chemicals

Dimethyl succinate, 98%, Thermo Scientific Chemicals

Catalog number: A12565.22
100 g, Each
Quantity
Catalog number: A12565.22
also known as A12565-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
106-65-0
IUPAC Name
1,4-dimethyl butanedioate
Molecular Formula
C6H10O4
InChI Key
MUXOBHXGJLMRAB-UHFFFAOYSA-N
SMILES
COC(=O)CCC(=O)OC
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥97.5% (UK-sourced material)
Assay from Supplier's CofA
≥97.5% (US-sourced material)
Comment
Material sourced in UK and US

Description

Dimethyl succinate is used as a flavoring agent. It have wide range of industrial applications such as Functional fluids (open systems), Intermediates, Paint additives and coating additives, Pigments Solvents, Viscosity adjustors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl succinate is used as a flavoring agent. It have wide range of industrial applications such as Functional fluids (open systems), Intermediates, Paint additives and coating additives, Pigments Solvents, Viscosity adjustors.

Solubility
Soluble in water. (122,9 g/L at 20°C). soluble in acetone, ethanol, very soluble in ethyl ether.

Notes
Store at room temperature. Ensure good ventilation.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text