5-Fluoro-2-nitrophenol, 98%, Thermo Scientific Chemicals
5-Fluoro-2-nitrophenol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Fluoro-2-nitrophenol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A12568.09
also known as A12568-09
Price (USD)/ Each
35.70
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
35.70
Add to cart
5-Fluoro-2-nitrophenol, 98%, Thermo Scientific Chemicals
Catalog numberA12568.09
Price (USD)/ Each
35.70
-
Add to cart
Chemical Identifiers
CAS446-36-6
IUPAC Name5-fluoro-2-nitrophenol
Molecular FormulaC6H4FNO3
InChI KeyQQURWFRNETXFTN-UHFFFAOYSA-N
SMILESOC1=CC(F)=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to dark yellow to orange to brown
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥97.5%
Melting Point (clear melt)30.0-37.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only