Thermo Scientific Chemicals

Hexamethylphosphorous triamide, 97%, Thermo Scientific Chemicals

Catalog number: A12571.18
50 g, Each
Thermo Scientific Chemicals

Hexamethylphosphorous triamide, 97%, Thermo Scientific Chemicals

Catalog number: A12571.18
50 g, Each
Quantity
Catalog number: A12571.18
also known as A12571-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1608-26-0
IUPAC Name
[bis(dimethylamino)phosphanyl]dimethylamine
Molecular Formula
C6H18N3P
InChI Key
XVDBWWRIXBMVJV-UHFFFAOYSA-N
SMILES
CN(C)P(N(C)C)N(C)C
Refractive Index
1.4630-1.4680 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms

Description

Hexamethylphosphorous triamide is used as a reagent in organic synthesis as a phosphorylating agent. It is associated with carbon tetrachloride for the substitution of hydroxy groups with chlorides. It is involved in the preparation of epoxides and arene oxides from aldehydes and aryldialdehdyes respectively. It is used in the preparation of carbonates as well as reduction of ozonides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethylphosphorous triamide is used as a reagent in organic synthesis as a phosphorylating agent. It is associated with carbon tetrachloride for the substitution of hydroxy groups with chlorides. It is involved in the preparation of epoxides and arene oxides from aldehydes and aryldialdehdyes respectively. It is used in the preparation of carbonates as well as reduction of ozonides.

Solubility
Miscible with water.

Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text