Thermo Scientific Chemicals

1-Methylimidazole, 99%, Thermo Scientific Chemicals

Catalog number: A12575.A3
2 kg, Each
Thermo Scientific Chemicals

1-Methylimidazole, 99%, Thermo Scientific Chemicals

Catalog number: A12575.A3
2 kg, Each
Quantity
Catalog number: A12575.A3
also known as A12575-A3
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
616-47-7
IUPAC Name
1-methyl-1H-imidazole
Molecular Formula
C4H6N2
InChI Key
MCTWTZJPVLRJOU-UHFFFAOYSA-N
SMILES
CN1C=CN=C1
Assay (GC)
≥98.5%
Comment
Purchased in the U.S. and in other countries
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Identification (FTIR)
Conforms (non-U.S. specification)

Description

1-Methylimidazole is used as a precursor for the synthesis of pyrrole-imidazole polyamides, ionic liquids such as 1-butyl-3-methylimidazolium hexafluorophosphate. It is actively involved in removing acid during the production of diethoxyphenylphosphine. It is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylimidazole is used as a precursor for the synthesis of pyrrole-imidazole polyamides, ionic liquids such as 1-butyl-3-methylimidazolium hexafluorophosphate. It is actively involved in removing acid during the production of diethoxyphenylphosphine. It is used as an intermediate in organic synthesis.

Solubility
Miscible with water.

Notes
Incompatible with carbon dioxide and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text