Thermo Scientific Chemicals

4-Fluorophenyl isocyanate, 98+%, Thermo Scientific Chemicals

Catalog number: A12576.14
25 g, Each
Thermo Scientific Chemicals

4-Fluorophenyl isocyanate, 98+%, Thermo Scientific Chemicals

Catalog number: A12576.14
25 g, Each
Quantity
Catalog number: A12576.14
also known as A12576-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1195-45-5
IUPAC Name
1-fluoro-4-isocyanatobenzene
Molecular Formula
C7H4FNO
InChI Key
DSVGFKBFFICWLZ-UHFFFAOYSA-N
SMILES
FC1=CC=C(C=C1)N=C=O
Assay (GC)
>98.0%
Refractive Index
1.5120 - 1.5160 @ 20?C
Form
Slightly hazy liquid
Comment
May form slight precipitate on storage
Appearance (Color)
Clear colorless to pale yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text