Thermo Scientific Chemicals

Phthalimide, 99%, Thermo Scientific Chemicals

Catalog number: A12584.0C
10,000 g, Each
Thermo Scientific Chemicals

Phthalimide, 99%, Thermo Scientific Chemicals

Catalog number: A12584.0C
10,000 g, Each
Quantity
Catalog number: A12584.0C
also known as A12584-0C
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
85-41-6
IUPAC Name
2,3-dihydro-1H-isoindole-1,3-dione
Molecular Formula
C8H5NO2
InChI Key
XKJCHHZQLQNZHY-UHFFFAOYSA-N
SMILES
O=C1NC(=O)C2=CC=CC=C12
Appearance (Color)
White to pale cream or pale yellow
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder or flakes
Assay (HPLC)
≥98.5%

Description

Phthalimide is a reagent used to transform allyl- and alkyl halides into protected primary amines. . It is used as a precursor to other organic compounds as a masked source of ammonia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthalimide is a reagent used to transform allyl- and alkyl halides into protected primary amines. . It is used as a precursor to other organic compounds as a masked source of ammonia.

Solubility
Slightly soluble in water. Soluble in aqueous alkali hydroxides

Notes
Store at room temperature. Ensure good ventilation.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text