Thermo Scientific Chemicals

Acetamide, 99%, Thermo Scientific Chemicals

Catalog number: A12589.0B
1000 g, Each
Thermo Scientific Chemicals

Acetamide, 99%, Thermo Scientific Chemicals

Catalog number: A12589.0B
1000 g, Each
Quantity
Catalog number: A12589.0B
also known as A12589-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
60-35-5
IUPAC Name
acetamide
Molecular Formula
C2H5NO
InChI Key
DLFVBJFMPXGRIB-UHFFFAOYSA-N
SMILES
CC(N)=O
Identification (FTIR)
Conforms
Melting Point (clear melt)
76.0-83.0?C
Appearance (Color)
Colorless to white
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.5%

Description

Acetamide is used as an industrial solvent, plasticizer, wetting and penetrating agent. It is also used for the transamidation of carboxamides in 1,4-dioxane. It finds application in lacquers and soldering flux. Further, it acts as a precursor to thioacetamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetamide is used as an industrial solvent, plasticizer, wetting and penetrating agent. It is also used for the transamidation of carboxamides in 1,4-dioxane. It finds application in lacquers and soldering flux. Further, it acts as a precursor to thioacetamide.

Solubility
Soluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text