Acetamide, 99%, Thermo Scientific Chemicals
Acetamide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetamide, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5000 g
250 g
1000 g
Catalog number A12589.0I
also known as A12589-0I
Price (USD)/ Each
405.65
Online exclusive
451.00 
Save 45.35 (10%)
-
Add to cart
Quantity:
5000 g
Request bulk or custom format
Price (USD)/ Each
405.65
Online exclusive
451.00 
Save 45.35 (10%)
Add to cart
Acetamide, 99%, Thermo Scientific Chemicals
Catalog numberA12589.0I
Price (USD)/ Each
405.65
Online exclusive
451.00 
Save 45.35 (10%)
-
Add to cart
Chemical Identifiers
CAS60-35-5
IUPAC Nameacetamide
Molecular FormulaC2H5NO
InChI KeyDLFVBJFMPXGRIB-UHFFFAOYSA-N
SMILESCC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Melting Point (clear melt)76.0-83.0?C
Appearance (Color)Colorless to white
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
View more
Acetamide is used as an industrial solvent, plasticizer, wetting and penetrating agent. It is also used for the transamidation of carboxamides in 1,4-dioxane. It finds application in lacquers and soldering flux. Further, it acts as a precursor to thioacetamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetamide is used as an industrial solvent, plasticizer, wetting and penetrating agent. It is also used for the transamidation of carboxamides in 1,4-dioxane. It finds application in lacquers and soldering flux. Further, it acts as a precursor to thioacetamide.

Solubility
Soluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Useful additive for brominations of acid-sensitive compounds in non-polar solvents since it forms a stable, insoluble 1:1 complex with HBr: Chem. Ber., 82, 275 (1949).
  2. For amidocarbonylation in the presence of carbon monoxide and hydrogen in a route to phenylalanine, see Benzyl chloride, A12481.
  3. D'Amico, F.; Rossi, B.; Camisasca, G.; Bencivenga, F.; Gessini, A.; Principi, E.; Cucini, R.; Masciovecchio, C. Slow-to-fast transition of hydrogen bond dynamics in acetamide hydration shell formation. Phys. Chem. Chem. Phys. 2015, 17 (16), 10987-10992.
  4. Aguilar-Pineda, J. A.; Méndez-Maldonado, G. A.; Núñez-Rojas, E.; Alejandre, J. Parametrisation of a force field of acetamide for simulations of the liquid phase. Mol. Phys. 2015, 113 (17-18), 2716-2724.