Thermo Scientific Chemicals

6-Aminobenzothiazole, 98+%, Thermo Scientific Chemicals

Catalog number: A12601.03
1 g, Each
Thermo Scientific Chemicals

6-Aminobenzothiazole, 98+%, Thermo Scientific Chemicals

Catalog number: A12601.03
1 g, Each
Quantity
Catalog number: A12601.03
also known as A12601-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
533-30-2
IUPAC Name
1,3-benzothiazol-6-amine
Molecular Formula
C7H6N2S
InChI Key
FAYAYUOZWYJNBD-UHFFFAOYSA-N
SMILES
NC1=CC=C2N=CSC2=C1
Appearance (Color)
Cream to dark cream to yellow to green to dark green to brown
Assay (GC)
≥98.0%
Melting Point (clear melt)
83.0-90.0?C
Form
Powder

Description

6-Aminobenzothiazole is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Aminobenzothiazole is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text