3-Bromobenzoic acid was used as test solute in the determination of acidity constants by capillary zone electrophoresis. It was used as internal standard to study the retention mechanisms of an unmodified and a hydroxylated polystyrene-divinylbenzene polymer by solid-phase extraction. It was also used in synthesis of N-(1,1-dimethyl-2-hydroxyethyl)-3-bromobenzamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Bromobenzoic acid was used as test solute in the determination of acidity constants by capillary zone electrophoresis. It was used as internal standard to study the retention mechanisms of an unmodified and a hydroxylated polystyrene-divinylbenzene polymer by solid-phase extraction. It was also used in synthesis of N-(1,1-dimethyl-2-hydroxyethyl)-3-bromobenzamide.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
- Joan Marc Cabot; Elisabet Fuguet; Clara Rafols; Marti Roses. Fast high-throughput method for the determination of acidity constants by capillary electrophoresis. II. Acidic internal standards. Journal of Chromatography A. 2010, 1217, (52)8340-8345
- Susanne Jonsso; Hans Boren. Analysis of mono- and diesters of o-phthalic acid by solid-phase extractions with polystyrene-divinylbenzene-based polymers. Journal of Chromatography A. 2002, 963, (1-2)393-400