Thermo Scientific Chemicals

2-Bromo-4'-chloroacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: A12607.14
25 g, Each
Thermo Scientific Chemicals

2-Bromo-4'-chloroacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: A12607.14
25 g, Each
Quantity
Catalog number: A12607.14
also known as A12607-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
536-38-9
IUPAC Name
2-bromo-1-(4-chlorophenyl)ethan-1-one
Molecular Formula
C8H6BrClO
InChI Key
FLAYZKKEOIAALB-UHFFFAOYSA-N
SMILES
ClC1=CC=C(C=C1)C(=O)CBr
Appearance (Color)
White to cream
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder or fused solid
Melting Point (clear melt)
93.0-100.0?C

Description

2-Bromo-4'-chloroacetophenone is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4′-chloroacetophenone is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text