Thermo Scientific Chemicals

Polytetrafluoroethylene, Thermo Scientific Chemicals

Catalog number: A12613.18
50 g, Each
Thermo Scientific Chemicals

Polytetrafluoroethylene, Thermo Scientific Chemicals

Catalog number: A12613.18
50 g, Each
Quantity
Catalog number: A12613.18
also known as A12613-18
Price (USD)
Have Questions?

Chemical Identifiers

CAS
9002-84-0
Molecular Formula
(C2F4)n
SMILES
FC(F)(-*)C(F)(F)-*
MDL Number
MFCD00084455
Appearance (Color)
White
Loss on Drying
≤1% (UK-sourced material)
Form
Powder
Comment
Material sourced in UK and US
Melting Point
327.5-337.5?C (US-sourced material)

Description

PTFE may be used as a non stick coating. It can be used as engineering plastics. It is employed as lining reaction vessels, gaskets and pump packing, electrical insulator for high frequency applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text