Methylphosphonic acid, 98%, Thermo Scientific Chemicals
Methylphosphonic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methylphosphonic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A12619.22
Price (USD)/ Each
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS993-13-5
IUPAC Namemethylphosphonic acid
Molecular FormulaCH5O3P
InChI KeyYACKEPLHDIMKIO-UHFFFAOYSA-N
SMILESCP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)103-109°C
Water Content (Karl Fischer Titration)≤1.5%
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder.
View more
Methylphosphonic acid is used to produce methyl-phosphonic acid dichloride by chlorination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylphosphonic acid is used to produce methyl-phosphonic acid dichloride by chlorination.

Solubility
Soluble in water and alcohol.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. Shao, X.; Ge, H.; Li, Z.; Ren, C.; Wang, J. Solubility of methylphosphonic acid in selected organic solvents. Fluid Phase Equilib. 2015, 390, 7-13.
  2. Rodin, I.; Baygildiev, T.; Stavrianidi, A.; Braun, A.; Rybalchenko, I.; Shpigun, O. Hydrophilic Interaction Liquid Chromatography Tandem Mass Spectrometry Methylphosphonic Acid Determination in Water Samples after Derivatization with p-Bromophenacyl Bromide. Chromatographia 2015, 78 (8), 585-591.