Tetra-n-butylammonium hydroxide, 40% w/w in methanol is used as a phase-transfer catalyst in organic synthesis especially for alkylation and condensation reactions. It is also used as a curing accelerator for epoxy resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetra-n-butylammonium hydroxide, 40% w/w in methanol is used as a phase-transfer catalyst in organic synthesis especially for alkylation and condensation reactions. It is also used as a curing accelerator for epoxy resins.
Solubility
Miscible with organic solvents.
Notes
Air sensitive and hygroscopic. Incompatible with strong acids and corrodes metal.
RUO – Research Use Only
Bauer, M.; Spange, S. Enhancing the Reactivity of an Electrophilic Barbiturate Dye by Cooperative Hydrogen Bonding. Eur. J. Org. Chem. 2010, 2010 (2), 259-264.
Clark, D. C.; Baker, W. E.; Russell, K. E.; Whitney, R. A. Dual monomer grafting of styrene and maleic anhydride onto model hydrocarbon substrates. J. Polym. Sci. A Polym. Chem. 2000, 38 (13), 2456-2468.