3-Methoxybenzyl chloride, 98+%, Thermo Scientific Chemicals
3-Methoxybenzyl chloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methoxybenzyl chloride, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
50 g
250 g
Catalog number A12630.14
also known as A12630-14
Price (USD)/ Each
103.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
103.00
Add to cart
3-Methoxybenzyl chloride, 98+%, Thermo Scientific Chemicals
Catalog numberA12630.14
Price (USD)/ Each
103.00
-
Add to cart
Chemical Identifiers
CAS824-98-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5420-1.5460 @ 20?C
FormLiquid
Identification (FTIR)Conforms
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only