Thermo Scientific Chemicals

(S)-(-)-1-Phenylethylamine, 98%, Thermo Scientific Chemicals

Catalog number: A12632.14
25 g, Each
Thermo Scientific Chemicals

(S)-(-)-1-Phenylethylamine, 98%, Thermo Scientific Chemicals

Catalog number: A12632.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A12632.14
also known as A12632-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2627-86-3
IUPAC Name
(1S)-1-phenylethan-1-amine
Molecular Formula
C8H11N
InChI Key
RQEUFEKYXDPUSK-ZETCQYMHSA-N
SMILES
C[C@H](N)C1=CC=CC=C1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Optical Rotation
-30 ? 2? (c=10 in ethanol) or -40? ± 1? (neat)
Assay (GC)
≥97.5%
Refractive Index
1.5240-1.5280 @ 20?C

Description

(S)-(-)-1-Phenylethylamine is utilized in a one-pot, multi-component synthesis of a highly substituted, chiral pyrrole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-1-Phenylethylamine is utilized in a one-pot, multi-component synthesis of a highly substituted, chiral pyrrole.

Solubility
Slightly miscible with water.

Notes
Keep away from sources of ignition. Incompatible with acids, acid chlorides, acid anhydrides and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text