Thermo Scientific Chemicals

4-Bromobiphenyl, 98+%, Thermo Scientific Chemicals

Catalog number: A12638.30
250 g, Each
Thermo Scientific Chemicals

4-Bromobiphenyl, 98+%, Thermo Scientific Chemicals

Catalog number: A12638.30
250 g, Each
Quantity
Catalog number: A12638.30
also known as A12638-30
Price (USD)
Have Questions?

Chemical Identifiers

CAS
92-66-0
IUPAC Name
4-bromo-1,1'-biphenyl
Molecular Formula
C12H9Br
InChI Key
PKJBWOWQJHHAHG-UHFFFAOYSA-N
SMILES
BrC1=CC=C(C=C1)C1=CC=CC=C1
Form
Crystals or powder or crystalline powder or flakes
Assay (GC)
≥98.0%
Melting Point (clear melt)
87.0-93.0?C
Appearance (Color)
White

Description

4-Bromobiphenyl is used as halogenated hydrocarbon. It is also used in used in the synthesis of 4-biphenylthiolate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobiphenyl is used as halogenated hydrocarbon. It is also used in used in the synthesis of 4-biphenylthiolate.

Solubility
Insoluble in water.

Notes
Sensitive to light. Store in cool dry place in tightly closed container. Avoid excessive heat and light. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text