2,2'-Methylenebis(4-chlorophenol), 95%, Thermo Scientific Chemicals
2,2'-Methylenebis(4-chlorophenol), 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2'-Methylenebis(4-chlorophenol), 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number A12640.36
Price (USD)/ Each
208.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
208.00
Add to cart
2,2'-Methylenebis(4-chlorophenol), 95%, Thermo Scientific Chemicals
Catalog numberA12640.36
Price (USD)/ Each
208.00
-
Add to cart
Chemical Identifiers
CAS97-23-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to pale pink
Assay (GC)≥94.0%
Melting Point (clear melt)170.5-179.5?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only