Thermo Scientific Chemicals

2-Ethylhexanoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A12644.AP
500 mL, Each
Thermo Scientific Chemicals

2-Ethylhexanoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A12644.AP
500 mL, Each
Quantity
Catalog number: A12644.AP
also known as A12644-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
149-57-5
IUPAC Name
2-ethylhexanoic acid
Molecular Formula
C8H16O2
InChI Key
OBETXYAYXDNJHR-UHFFFAOYNA-N
SMILES
CCCCC(CC)C(O)=O
Form
Liquid
Refractive Index
1.4235-1.4265 @ 20?C
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.5%

Description

2-Ethylhexanoic acid is used in the preparation of metal derivatives, which act as a catalyst in polymerization reactions. For example, tin 2-ethylhexanoate is used in the manufacturing of poly(lactic-co-glycolic acid). It is also used as a stabilizer for polyvinyl chlorides. It is also involved in solvent extraction and dye granulation. Further, it is used to prepare plasticizers, lubricants, detergents, flotation aids, corrosion inhibitors and alkyd resins. In addition to this, it serves as a catalyst for polyurethane foaming.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylhexanoic acid is used in the preparation of metal derivatives, which act as a catalyst in polymerization reactions. For example, tin 2-ethylhexanoate is used in the manufacturing of poly(lactic-co-glycolic acid). It is also used as a stabilizer for polyvinyl chlorides. It is also involved in solvent extraction and dye granulation. Further, it is used to prepare plasticizers, lubricants, detergents, flotation aids, corrosion inhibitors and alkyd resins. In addition to this, it serves as a catalyst for polyurethane foaming.

Solubility
Miscible with water.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text