Thermo Scientific Chemicals

Triethylamine, 99%, Thermo Scientific Chemicals

Catalog number: A12646.AP
500 mL, Each
Thermo Scientific Chemicals

Triethylamine, 99%, Thermo Scientific Chemicals

Catalog number: A12646.AP
500 mL, Each
Quantity
Catalog number: A12646.AP
also known as A12646-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
121-44-8
IUPAC Name
triethylamine
Molecular Formula
C6H15N
InChI Key
ZMANZCXQSJIPKH-UHFFFAOYSA-N
SMILES
CCN(CC)CC
Comment
Specification differs for U.S. and non-U.S. material where indicated
Refractive Index
1.3990-1.4020 @ 20°C (non-U.S. specification)
Form
Liquid
Identification (FTIR)
Conforms (non-U.S. specification)
Appearance (Color)
Clear colorless to very pale yellow

Description

Triethylamine is a base used to prepare esters and amides from acyl chlorides as well as in the synthesis of quaternary ammonium compounds. It acts as a catalyst in the formation of urethane foams and epoxy resins, dehydrohalogeantion reactions, acid neutralizer for condensation reactions and Swern oxidations. It finds application in reverse phase high-performance liquid chromatography (HPLC) as a mobile-phase modifier. It is also used as an accelerator activator for rubber, as a propellant, as a corrosion inhibitor, as a curing and hardening agent for polymers and for the desalination of seawater. Furthermore, it is used in automotive casting industry and textile industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethylamine is a base used to prepare esters and amides from acyl chlorides as well as in the synthesis of quaternary ammonium compounds. It acts as a catalyst in the formation of urethane foams and epoxy resins, dehydrohalogeantion reactions, acid neutralizer for condensation reactions and Swern oxidations. It finds application in reverse phase high-performance liquid chromatography (HPLC) as a mobile-phase modifier. It is also used as an accelerator activator for rubber, as a propellant, as a corrosion inhibitor, as a curing and hardening agent for polymers and for the desalination of seawater. Furthermore, it is used in automotive casting industry and textile industry.

Solubility
Miscible with water, ether and ethanol.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text