Sodium 1-butanesulfonate was used in the synthesis of high silicon content SAPO4-5. It was also used as a mobile phase to separate eight selenium compounds (selenite, selenate, selenocystine, selenourea, selenomethionine, selenoethionine, selenocystamine and trimethylselenonium ion). Ion-associating reagent for HPLC1 and reversed-phase chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium 1-butanesulfonate was used in the synthesis of high silicon content SAPO4-5. It was also used as a mobile phase to separate eight selenium compounds (selenite, selenate, selenocystine, selenourea, selenomethionine, selenoethionine, selenocystamine and trimethylselenonium ion). Ion-associating reagent for HPLC1 and reversed-phase chromatography.
Solubility
Soluble in water (1-10%).
Notes
Sensitive to light. Store in cool dry place in tightly closed container. Avoid excessive heat and light. Store away from oxidizing agent.
RUO – Research Use Only
General References:
- Xue S Han et. al. Synthesis of high silicon content SAPO(4)-5 using anionic surfactants in a hexanol/aqueous two phase media. Chemical Communications. 2002, (2), 166-167.
- J Zheng, M Ohata; N Furuta; W Kosmus; Jian Zheng; Masaki Ohata; Naoki Furuta; Walter Kosmusl. Speciation of selenium compounds with ion-pair reversed-phase liquid chromatography using inductively coupled plasma mass spectrometry as element-specific detection. Journal of Chromatography A. 2000, 874,(1), 55-64.