Thermo Scientific Chemicals

1-Phenyl-1H-tetrazole-5-thiol, 99%, Thermo Scientific Chemicals

Catalog number: A12656.36
500 g, Each
Thermo Scientific Chemicals

1-Phenyl-1H-tetrazole-5-thiol, 99%, Thermo Scientific Chemicals

Catalog number: A12656.36
500 g, Each
Quantity
Catalog number: A12656.36
also known as A12656-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
86-93-1
IUPAC Name
1-phenyl-2,5-dihydro-1H-1,2,3,4-tetrazole-5-thione
Molecular Formula
C7H6N4S
InChI Key
GGZHVNZHFYCSEV-UHFFFAOYSA-N
SMILES
S=C1N=NNN1C1=CC=CC=C1
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥98.5%
Identification (FTIR)
Conforms

Description

1-Phenyl-1H-tetrazole-5-thiol is an effective inhibitor of aluminum corrosion in 1M HCl solution. 1-Phenyl-1H-tetrazole-5-thiol was used in the synthesis of oxacyclic building blocks via highly stereoselective radical cyclization and olefin metathesis reactions. It was also used in the synthesis of metalated tetradecyl sulfone. It is also used in the spectrophotometric determination of Pt and Bi.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenyl-1H-tetrazole-5-thiol is an effective inhibitor of aluminum corrosion in 1M HCl solution. 1-Phenyl-1H-tetrazole-5-thiol was used in the synthesis of oxacyclic building blocks via highly stereoselective radical cyclization and olefin metathesis reactions. It was also used in the synthesis of metalated tetradecyl sulfone. It is also used in the spectrophotometric determination of Pt and Bi.

Solubility
Soluble in 5% ethanol, water (partly), acetone, chloroform, and methanol.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text