Thermo Scientific Chemicals

5-Methylbenzo[b]thiophene, 97%, Thermo Scientific Chemicals

Catalog number: A12666.03
1 g, Each
Thermo Scientific Chemicals

5-Methylbenzo[b]thiophene, 97%, Thermo Scientific Chemicals

Catalog number: A12666.03
1 g, Each
Quantity
Catalog number: A12666.03
also known as A12666-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14315-14-1
IUPAC Name
5-methyl-1-benzothiophene
Molecular Formula
C9H8S
InChI Key
DOHZWDWNQFZIKH-UHFFFAOYSA-N
SMILES
CC1=CC=C2SC=CC2=C1
Form
crystals or fused solid
Appearance (Color)
White to yellow to pale brown
Comment
Material darkens on storage
Assay (GC)
>96%

Description

5-Methylbenzo[b]thiophene is useful in substituted thiophene for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methylbenzo[b]thiophene is useful in substituted thiophene for proteomics research.

Solubility
Insoluble in water.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text