2',3'-Dichloroacetophenone, 98%, Thermo Scientific Chemicals
2',3'-Dichloroacetophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2',3'-Dichloroacetophenone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A12667.03
also known as A12667-03
Price (USD)/ Each
41.50
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
41.50
Add to cart
2',3'-Dichloroacetophenone, 98%, Thermo Scientific Chemicals
Catalog numberA12667.03
Price (USD)/ Each
41.50
-
Add to cart
Chemical Identifiers
CAS56041-57-7
IUPAC Name1-(2,3-dichlorophenyl)ethan-1-one
Molecular FormulaC8H6Cl2O
InChI KeyKMABBMYSEVZARZ-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC(Cl)=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive Index1.5585-1.5625 @ 20?C
Assay (GC)≥97.5%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only