Thermo Scientific Chemicals

4-Aminobenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A12673.0B
1000 g, Each
Thermo Scientific Chemicals

4-Aminobenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A12673.0B
1000 g, Each
Quantity
Catalog number: A12673.0B
also known as A12673-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
150-13-0
IUPAC Name
4-aminobenzoic acid
Molecular Formula
C7H7NO2
InChI Key
ALYNCZNDIQEVRV-UHFFFAOYSA-N
SMILES
NC1=CC=C(C=C1)C(O)=O
Form
Powder
Assay (Silylated GC)
≥98.5
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Identification (FTIR)
Conforms
Appearance (Color)
White to pale brown

Description

4-Aminobenzoic acid acts as a gastric acid secretion inhibitor. It is employed as an intermediate in the synthesis of folate (Vitamin BX), azo dyes and cross linking agents. It is also useful in biomedical sector for the preparation of local anesthetic and ointments. It is widely used in industries to make pharmaceutical products, flavors and preservatives. It is actively involved as a UV-blocking ingredient in sun tan cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminobenzoic acid acts as a gastric acid secretion inhibitor. It is employed as an intermediate in the synthesis of folate (Vitamin BX), azo dyes and cross linking agents. It is also useful in biomedical sector for the preparation of local anesthetic and ointments. It is widely used in industries to make pharmaceutical products, flavors and preservatives. It is actively involved as a UV-blocking ingredient in sun tan cosmetics.

Solubility
Slightly soluble in water. Soluble in methanol and diethyl ether.

Notes
Air and light sensitive. Incompatible with ferric salts and oxidizing agents. May discolor on exposure to light and air. Keep the container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text