3-Amino-4-methylbenzoic acid, is a raw material used in synthesis. It is also used as an organic intermediate, Pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Amino-4-methylbenzoic acid, is a raw material used in synthesis. It is also used as an organic intermediate, Pharmaceutical intermediate.
Solubility
Very soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperature and pressures.
RUO – Research Use Only
General References:
- MJS Monte.; DM Hillesheim. Thermodynamic study of the sublimation of six aminomethylbenzoic acids. The Journal of Chemical Thermodynamics. 2001, 33, (7), 745-754.
- H Wada.; T Ishizuki.; G Nakagawa. 2-(2-thiazolylazo)-4-methyl-5-(sulfomethylamino) benzoic acid as a reagent for the spectrophotometric determination of cobalt. Analytica Chimica Acta. 1982, 1 35, (2), 333-341.