Thermo Scientific Chemicals

3-Amino-4-methylbenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A12674.14
25 g, Each
Thermo Scientific Chemicals

3-Amino-4-methylbenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A12674.14
25 g, Each
Quantity
Catalog number: A12674.14
also known as A12674-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2458-12-0
IUPAC Name
3-amino-4-methylbenzoic acid
Molecular Formula
C8H9NO2
InChI Key
XKFIFYROMAAUDL-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1N)C(O)=O
Appearance (Color)
Pale cream to cream to brown
Melting Point (clear melt)
165.0-171.0?C
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥98.5%
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

3-Amino-4-methylbenzoic acid, is a raw material used in synthesis. It is also used as an organic intermediate, Pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-4-methylbenzoic acid, is a raw material used in synthesis. It is also used as an organic intermediate, Pharmaceutical intermediate.

Solubility
Very soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperature and pressures.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text