2-Methylstyrene, 98%, stab. with 0.1% 4-tert-butylcatechol, Thermo Scientific Chemicals
2-Methylstyrene, 98%, stab. with 0.1% 4-tert-butylcatechol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methylstyrene, 98%, stab. with 0.1% 4-tert-butylcatechol, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A12676.14
also known as A12676-14
Price (USD)/ Each
607.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
607.00
Add to cart
2-Methylstyrene, 98%, stab. with 0.1% 4-tert-butylcatechol, Thermo Scientific Chemicals
Catalog numberA12676.14
Price (USD)/ Each
607.00
-
Add to cart
Chemical Identifiers
CAS611-15-4
IUPAC Name1-ethenyl-2-methylbenzene
Molecular FormulaC9H10
InChI KeyNVZWEEGUWXZOKI-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Solution TestClear: 10% in Diethyl ether
Appearance (Color)Clear colorless to pale yellow
Solution TestClear: 10% in Methanol
FormLiquid
Refractive Index1.5410-1.5460 @ 20?C
View more
2-Methylstyrene is a monomer used in the preparation of poly(2-methylstyrene). It is also used as an intermediate in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylstyrene is a monomer used in the preparation of poly(2-methylstyrene). It is also used as an intermediate in synthetic chemistry.

Solubility
Slightly miscible with water.

Notes
Store in a cool place. Incompatible with oxidizing agents and strong acids.
RUO – Research Use Only

General References:

  1. Wang, M.; Gao, B.; Huang, H. Catalytic nucleophilic addition of olefinic C-H bond to alpha, beta-unsaturated-gama-lactams. Tetrahedron Lett. 2015, 56 (41), 5533-5536.
  2. Chen, J.; Hua, L.; Chen, C.; Guo, L.; Zhang, R.; Chen, A.; Xiu, Y.; Liu, X.; Hou, Z. Temperature-Dependent Immobilization of a Tungsten Peroxo Complex That Catalyzes the Hydroxymethoxylation of Olefins. ChemPlusChem 2015, 80 (6), 1029-1037.