4-Amino-1-butanol is a useful intermediate in a variety of organic synthesis. It is mainly used in personal-care products, preparation of water soluble cationic flocculants and ion exchange resins as well as in the preparation of beta-lactam antibiotics. It is used for the production of efficient anionic emulsifiers, nonionic polyethylene emulsions, water treatment, metal treatment and absorption of carbon dioxide gas. It is used as a pigment dispersion aid and curing agent in selected textile -resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Amino-1-butanol is a useful intermediate in a variety of organic synthesis. It is mainly used in personal-care products, preparation of water soluble cationic flocculants and ion exchange resins as well as in the preparation of beta-lactam antibiotics. It is used for the production of efficient anionic emulsifiers, nonionic polyethylene emulsions, water treatment, metal treatment and absorption of carbon dioxide gas. It is used as a pigment dispersion aid and curing agent in selected textile -resins.
Solubility
Miscible with water.
Notes
Air Sensitive and hygroscopic. Incompatible with strong oxidizing agents and acids.
RUO – Research Use Only
General References:
- Idris, Z.; Peresunko, N. P.; Jens, K. J.; Eimer, D. A. Equilibrium solubility of carbon dioxide in aqueous solutions of 3-amino-1-propanol, 4-amino-1-butanol and 5-amino-1-pentanol at low partial pressures. Fluid Phase Equilib. 2015, 387, 81-87.
- Ruan, Y.; Chen, S.; Brown, J. D.; Hadad, C. M.; Badjic, J. D. Ubiquitous Assembly of Amphiphilic Baskets into Unilamellar Vesicles and Their Recognition Characteristics. Org. Lett. 2015, 17 (4), 852-855.