Thermo Scientific Chemicals

4-Amino-1-butanol, 98%, Thermo Scientific Chemicals

Catalog number: A12680.14
25 g, Each
Thermo Scientific Chemicals

4-Amino-1-butanol, 98%, Thermo Scientific Chemicals

Catalog number: A12680.14
25 g, Each
Quantity
Catalog number: A12680.14
also known as A12680-14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13325-10-5
IUPAC Name
4-hydroxybutan-1-aminium
Molecular Formula
C4H12NO
InChI Key
BLFRQYKZFKYQLO-UHFFFAOYSA-O
SMILES
[NH3+]CCCCO
Form
Liquid
Water Content (Karl Fischer Titration)
≤1%
Refractive Index
1.4600-1.4640 @ 20?C
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless to pale yellow

Description

4-Amino-1-butanol is a useful intermediate in a variety of organic synthesis. It is mainly used in personal-care products, preparation of water soluble cationic flocculants and ion exchange resins as well as in the preparation of beta-lactam antibiotics. It is used for the production of efficient anionic emulsifiers, nonionic polyethylene emulsions, water treatment, metal treatment and absorption of carbon dioxide gas. It is used as a pigment dispersion aid and curing agent in selected textile -resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-1-butanol is a useful intermediate in a variety of organic synthesis. It is mainly used in personal-care products, preparation of water soluble cationic flocculants and ion exchange resins as well as in the preparation of beta-lactam antibiotics. It is used for the production of efficient anionic emulsifiers, nonionic polyethylene emulsions, water treatment, metal treatment and absorption of carbon dioxide gas. It is used as a pigment dispersion aid and curing agent in selected textile -resins.

Solubility
Miscible with water.

Notes
Air Sensitive and hygroscopic. Incompatible with strong oxidizing agents and acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text