2,3,5,6-Tetrafluorophenol is used as a cytochrome P450 substrate. 2,3,5,6-Tetrafluorophenol was used in preparation of radioiodinated phenylalanine derivative which is useful in peptide synthesis, technetium-99m labeled antibodies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3,5,6-Tetrafluorophenol is used as a cytochrome P450 substrate. 2,3,5,6-Tetrafluorophenol was used in preparation of radioiodinated phenylalanine derivative which is useful in peptide synthesis, technetium-99m labeled antibodies.
Solubility
Partly miscible with water.
Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
RUO – Research Use Only
General References:
- Howard B. Gamper; Michael W. Ree; Thomas Cox; Jeanne S. Virosco; A. David Adams; Alexander A. Gall; John K. Scholler; and Rich B. Meyer Jr. Facile preparation of nuclease resistant 3' modified oligodeoxynucleotides. Nucl. Acids Res. 1993, 21(1), 145-150.
- J.P. Maier; O. Marthaler; M. Mohraz. Emission spectra of the cations of some fluoro-substituted phenols in the gaseous phase. Journal of Electron Spectroscopy and Related Phenomena. 1980, 19(1), 11-20.