Thermo Scientific Chemicals

2-Acetylpyridine, 98%, Thermo Scientific Chemicals

Catalog number: A12688.36
500 g, Each
Thermo Scientific Chemicals

2-Acetylpyridine, 98%, Thermo Scientific Chemicals

Catalog number: A12688.36
500 g, Each
Quantity
Catalog number: A12688.36
also known as A12688-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1122-62-9
IUPAC Name
1-(pyridin-2-yl)ethan-1-one
Molecular Formula
C7H7NO
InChI Key
AJKVQEKCUACUMD-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=CC=N1
Assay (GC)
> 97.5%
Refractive Index
1.5220 - 1.5240 @20?C
Appearance (Color)
Clear, colourless to pale yellow
Form
liquid

Description

2-Acetylpyridine used as flavoring agent. It is also used in application for food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetylpyridine used as flavoring agent. It is also used in application for food additive.

Solubility
Soluble in water (18.2 g/100g @ 25C). Soluble in and acetate. Slightly soluble in carbon tetrachloride.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text