Thermo Scientific Chemicals

trans-2-Pentenoic acid, 90+%, remainder other isomers, Thermo Scientific Chemicals

Catalog number: A12689.09
10 g, Each
Thermo Scientific Chemicals

trans-2-Pentenoic acid, 90+%, remainder other isomers, Thermo Scientific Chemicals

Catalog number: A12689.09
10 g, Each
Quantity
Catalog number: A12689.09
also known as A12689-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13991-37-2
IUPAC Name
(2E)-pent-2-enoic acid
Molecular Formula
C5H8O2
InChI Key
YIYBQIKDCADOSF-ONEGZZNKSA-N
SMILES
CC\C=C\C(O)=O
Appearance (Color)
Clear, colourless to pale yellow
Assay (GC)
>90.0% (Silyl derivative; major isomer)
Form
liquid
Assay (Aqueous acid-base Titration)
>98.0%
Refractive Index
1.4495-1.4535

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text