Thermo Scientific Chemicals

Hippuric acid, 98%, Thermo Scientific Chemicals

Catalog number: A12690.0B
1000 g, Each
Thermo Scientific Chemicals

Hippuric acid, 98%, Thermo Scientific Chemicals

Catalog number: A12690.0B
1000 g, Each
Quantity
Catalog number: A12690.0B
also known as A12690-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
495-69-2
IUPAC Name
2-(phenylformamido)acetic acid
Molecular Formula
C9H9NO3
InChI Key
QIAFMBKCNZACKA-UHFFFAOYSA-N
SMILES
OC(=O)CNC(=O)C1=CC=CC=C1
Identification (FTIR)
Conforms
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Melting Point (clear melt)
187.0-194.0?C
Form
Crystals or powder or crystalline powder

Description

Hippuric acid is used as a intermediate for the manufacturing medicine and other organic compounds. Hippuric acid can be used to study cell biology, chemical biology, bioactive small molecules, amino acid derivatives, peptide synthesis, chemical synthesis and nutrition. Hippuric acid has been used to inform the metabolism and urinary excretion of procyanidins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hippuric acid is used as a intermediate for the manufacturing medicine and other organic compounds. Hippuric acid can be used to study cell biology, chemical biology, bioactive small molecules, amino acid derivatives, peptide synthesis, chemical synthesis and nutrition. Hippuric acid has been used to inform the metabolism and urinary excretion of procyanidins.

Solubility
Soluble in water.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text